Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylphenethylamine, 99%

Catalog No. p-3779179
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127450050 5 g Glass bottle
AC127450250 25 g Glass bottle
AC127451000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC127450050 Supplier Thermo Scientific Chemicals Supplier No. 127450050
Only null left
Add to Cart

CAS: 589-08-2 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-08-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008291
InChI Key SASNBVQSOZSTPD-UHFFFAOYSA-N
Synonym n-methylphenethylamine, benzeneethanamine, n-methyl, methyl-phenethyl-amine, n-methyl-2-phenylethylamine, n-methyl-n-2-phenylethyl amine, 1-phenyl-2-methylaminoethane, phenethylamine, n-methyl, methyl 2-phenylethyl amine, 1-phenyl-2-methylamino-aethan, n-methyl-phenethylamine
PubChem CID 11503
IUPAC Name N-methyl-2-phenylethanamine
SMILES CNCCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 203.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5150 to 1.5170
Linear Formula C6H5CH2CH2NHCH3
Quantity 5 g
Beilstein 12, 1097
Specific Gravity 0.93
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in alcohol and ether
Formula Weight 135.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylphenethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 209-632-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.