Learn More
Medchemexpress LLC N-Methylpiperazine-d11 | 1319723-22-2 | C₅HD₁₁N₂ | 50 MG
SDP
Catalog No. 5000432063
Encompass_Preferred
N-Methylpiperazine-d11 is the deuterium labeled N-Methylpiperazine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration is noted for its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32063 Supplier Medchemexpress LLC Supplier No. HY78871S150MG
N-Methylpiperazine-d11 is the deuterium labeled N-Methylpiperazine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration is noted for its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS