Learn More
Medchemexpress LLC N-methylpiperazine-d3 hydrochloride | 2043778-87-4 | 97.0% | 176.10 | C5H11D3Cl2N2 | 10 MG
SDP
Catalog No. 5000299280
Encompass_Preferred
N-Methylpiperazine-d3 hydrochloride is a deuterium-labeled hydrochloride salt of N-methylpiperazine supplied for research and analytical applications. It is provided as a high-purity isotopically labeled reference material intended for use as a tracer or internal standard in mass spectrometry and quantitative analyses.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Purity 97.0% d3, suitable for analytical work.
  • White to off-white solid with defined storage requirements to preserve integrity.
  • Stable when stored sealed at 4°C, away from moisture and light.
Catalog No. 50-002-99280 Supplier Medchemexpress LLC Supplier No. HY78871S610MG
N-Methylpiperazine-d3 hydrochloride is a deuterium-labeled hydrochloride salt of N-methylpiperazine supplied for research and analytical applications. It is provided as a high-purity isotopically labeled reference material intended for use as a tracer or internal standard in mass spectrometry and quantitative analyses.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Purity 97.0% d3, suitable for analytical work.
  • White to off-white solid with defined storage requirements to preserve integrity.
  • Stable when stored sealed at 4°C, away from moisture and light.