Learn More
Medchemexpress LLC N-Methylpiperazine-d3 hydrochloride | 2043778-87-4 | 97.0% | C5H11D3Cl2N2 | 100 MG
SDP
Catalog No. 5000434888
Encompass_Preferred
N-Methylpiperazine-d3 (hydrochloride) is a deuterated labeled N-Methylpiperazine, for research use only and not sold to patients. Deuteration, the substitution of hydrogen with deuterium, has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34888 Supplier Medchemexpress LLC Supplier No. HY78871S6100MG
N-Methylpiperazine-d3 (hydrochloride) is a deuterated labeled N-Methylpiperazine, for research use only and not sold to patients. Deuteration, the substitution of hydrogen with deuterium, has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS