Learn More
Medchemexpress LLC N-Methylpiperazine-d3 hydrochloride | 2043778-87-4 | 97.0% | C5H11D3Cl2N2 | 25 MG
SDP
Catalog No. 5000359019
Encompass_Preferred
N-Methylpiperazine-d3 hydrochloride is a deuterated labeled N-Methylpiperazine. Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration can affect pharmacokinetic and metabolic profiles. This compound is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes
  • Has potential to affect pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-003-59019 Supplier Medchemexpress LLC Supplier No. 25MGHY78871S6
N-Methylpiperazine-d3 hydrochloride is a deuterated labeled N-Methylpiperazine. Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration can affect pharmacokinetic and metabolic profiles. This compound is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes
  • Has potential to affect pharmacokinetic and metabolic profiles of drugs
  • For research use only