Learn More
Medchemexpress LLC N-Methylpiperazine-d3 hydrochloride | 2043778-87-4 | 97.0% | C5H11D3Cl2N2 | 50 MG
SDP
Catalog No. 5000432995
Encompass_Preferred
N-Methylpiperazine-d3 hydrochloride is a deuterated labeled N-Methylpiperazine, primarily intended for research use. This compound serves as a valuable tracer and can be utilized as an internal standard for various quantitative analytical techniques, including NMR, GC-MS, or LC-MS. For optimal preservation, store at 4°C in a sealed container, protected from moisture and light.
  • Can be used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32995 Supplier Medchemexpress LLC Supplier No. HY78871S650MG
N-Methylpiperazine-d3 hydrochloride is a deuterated labeled N-Methylpiperazine, primarily intended for research use. This compound serves as a valuable tracer and can be utilized as an internal standard for various quantitative analytical techniques, including NMR, GC-MS, or LC-MS. For optimal preservation, store at 4°C in a sealed container, protected from moisture and light.
  • Can be used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.