Learn More
Medchemexpress LLC N-Methylpiperazine-d4 | 343864-02-8 | MFCD03428306 | 99.4% | C5H8D4N2 | 1 MG
SDP
Catalog No. 5000434867
Encompass_Preferred
N-Methylpiperazine-d4 is the deuterium-labeled N-Methylpiperazine, intended for research use only. This compound serves as a tracer or internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Stable heavy isotopes, such as deuterium, are integrated into drug molecules primarily for quantitation during the drug development process. Deuteration is gaining recognition for its potential to influence the pharmacokinetic and metabolic profiles of various drugs.
  • Research-grade deuterium-labeled compound
  • Suitable for quantitative analysis
  • Can be used as a tracer or internal standard
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-34867 Supplier Medchemexpress LLC Supplier No. HY78871S21MG
N-Methylpiperazine-d4 is the deuterium-labeled N-Methylpiperazine, intended for research use only. This compound serves as a tracer or internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Stable heavy isotopes, such as deuterium, are integrated into drug molecules primarily for quantitation during the drug development process. Deuteration is gaining recognition for its potential to influence the pharmacokinetic and metabolic profiles of various drugs.
  • Research-grade deuterium-labeled compound
  • Suitable for quantitative analysis
  • Can be used as a tracer or internal standard
  • Potential to affect pharmacokinetic and metabolic profiles of drugs