Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methylpiperazine-d8 hydrochloride | 917358-65-7 | MFCD03428101 | 95.0% | 181.13 g/mol | C5H6D8Cl2N2 | 10 MG
SDP

Catalog No. 5000298502
Encompass_Preferred

N-Methylpiperazine-d8 hydrochloride is the deuterium-labeled isotopologue of N-methylpiperazine, supplied as the hydrochloride/dihydrochloride salt for research use. It is intended for analytical and synthetic applications where isotopic labeling is required, such as mass spectrometry internal standards, NMR studies, and metabolic tracing.

  • Deuterium-labeled (d8) isotopologue.
  • Suitable as an internal standard for mass spectrometry quantitation.
  • Applicable to NMR and metabolic tracing experiments.
  • Provided as a hydrochloride/dihydrochloride salt for improved stability and handling.
  • Typical vendor-reported purity approximately 95.0%.
  • Molecular weight 181.13 g/mol; chemical formula C5H6D8Cl2N2.
  • Supplied in small milligram quantities for analytical workflows.

Catalog No. 50-002-98502 Supplier Medchemexpress LLC Supplier No. HY78871S310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Methylpiperazine-d8 hydrochloride is the deuterium-labeled isotopologue of N-methylpiperazine, supplied as the hydrochloride/dihydrochloride salt for research use. It is intended for analytical and synthetic applications where isotopic labeling is required, such as mass spectrometry internal standards, NMR studies, and metabolic tracing.

  • Deuterium-labeled (d8) isotopologue.
  • Suitable as an internal standard for mass spectrometry quantitation.
  • Applicable to NMR and metabolic tracing experiments.
  • Provided as a hydrochloride/dihydrochloride salt for improved stability and handling.
  • Typical vendor-reported purity approximately 95.0%.
  • Molecular weight 181.13 g/mol; chemical formula C5H6D8Cl2N2.
  • Supplied in small milligram quantities for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.