Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpropionamide 99%, Thermo Scientific™

Catalog No. AC414800050
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC414800050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414800050 Supplier Thermo Scientific Chemicals Supplier No. 414800050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1187-58-2
CAS Min % 98.5
CAS Max % 100.0
Density 1.01
Infrared Spectrum Authentic
Molecular Formula C4H9NO
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
Linear Formula CH3CH2CONHCH3
Quantity 5g
MDL Number MFCD00009300
Synonym n-methylpropionamide, propanamide, n-methyl, propionamide, n-methyl, n-methylpropionic acid amide, methyl propionic acid amide, n-methylpropionsaureamid, unii-2tsz8hw6dq, n-methylpropionsaureamid german, 2tsz8hw6dq, n-methyl propionamide
InChI Key QJQAMHYHNCADNR-UHFFFAOYSA-N
SMILES CCC(=O)NC
IUPAC Name N-methylpropanamide
Molecular Weight (g/mol) 87.122
PubChem CID 14470
Formula Weight 87.12
Percent Purity ≥98.5%
Chemical Name or Material N-Methylpropionamide, 99%
Show More Show Less
EINECSNumber 214-699-7
RTECSNumber UE4515000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.