Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Methylpyrrolidine

Catalog No. M792045ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M792045ML 5 mL
M79204100ML 100 mL
M79204500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M792045ML Supplier Sigma Aldrich Supplier No. M792045ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-methylpyrrolidine can be used as a starting material to synthesize: n-methyl pyrrolidine-zinc borohydride (zbhnmp), a reducing agent for reduction of aldehydes, ketones, acid chlorides, and esters
  • Zbhnmp is also used in the reductive amination of aldehydes and ketones to their corresponding amines.n-alkyl-n-methyl-pyrrolidinium bis(trifluoromethanesulfonyl)imide ionic liquids.n-methylpyrrolidine-2-one hydrotribromide applicable as a catalyst for aziridination of alkenes

Specifications

CAS 120-94-5
Density 0.819 g/mL (at 25°C (literature))
Boiling Point 80°C to 81°C (lit.)
Molecular Formula C5H11N
Linear Formula C5H11N
MDL Number MFCD00003173
Quantity 5 mL
Molecular Weight (g/mol) 85.15
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.