Learn More
Description
Specifications
Specifications
| pH | 7.7 to 8.0 |
| Melting Point | -24°C |
| Absorbance | 0.02 AU max. at 400nm, 0.3 AU max. at 300nm, 0.7 AU max. at 270nm, 1.0 AU max. at 265nm |
| Color | Colorless |
| Density | 1.030g/cm³ |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Assay Percent Range | (by GC): ≥99.8% |
| Titratable Base | 0.001mEq/g max. |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Combustible liquid. Developmental hazard. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Obtain medical attention. NFPA Health:2 Flammability:2 Instability:0 |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.