Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpyrrolidone, for HPLC, GC and spectrophotometry, >99.5%, Solstice

Catalog No. 6002274 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-022-74 4 x 4 L Glass Bottle
60-022-70 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-022-74 Supplier Solstice Supplier No. 3044
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Absorbance 0.5 AU max. at 300 nm, 0.1 AU max. at 325 nm, 0.03 AU max. at 350 nm, 0.01 AU max. at 400 nm
CAS Max % 100.0000%
Density 1.0259 g/cm3
Boiling Point 202°C
Flash Point 88°C
Packaging Glass Bottle
Quantity 4 x 4 L
Beilstein 106420
Formula Weight 99.15
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material N-Methylpyrrolidone, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.