Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpyrrolidone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017873 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7873 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-7873 Supplier MilliporeSigma Supelco Supplier No. PHR13522G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 518°F
CAS 872-50-4
Vapor Pressure 0.29 mmHg (20°C)
Vapor Density 3.4 (vs. air)
Density 1.028 g/mL (at 25°C)
Flash Point 91°C - Closed Cup
Physical Form Neat
Linear Formula C5H9 NO
MDL Number MFCD00003193
Melting Point -24°C
Boiling Point 202°C; 81°C to 82°C (10 mmHg)
Synonym 1-Methyl-2-pyrrolidinone; 1-Methyl-2-pyrrolidone; N-Methyl-2-pyrrolidone; NMP
Type Certified Reference Material
Quantity 2 g
Formula Weight 99.13
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.