Learn More
Selleck Chemical LLC N-Methylsarcosine
SDP
Catalog No. 5000004235
Encompass_Preferred
N-Methylsarcosine (N N-Dimethylglycine (Dimethylamino)acetic acid) is a natural N-methylated glycine that is used in the development of glycine-based ionic liquids and emulsifiers
Catalog No. 50-000-04235 Supplier Selleck Chemical LLC Supplier No. S605625MG
N-Methylsarcosine (N N-Dimethylglycine (Dimethylamino)acetic acid) is a natural N-methylated glycine that is used in the development of glycine-based ionic liquids and emulsifiers