Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylsulfanilic acid, 98+%, Thermo Scientific™

Catalog No. AAL0101614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0101614 25 g
AAL0101606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0101614 Supplier Thermo Scientific Chemicals Supplier No. L0101614

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24447-99-2
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00014732
InChI Key QRAXZXPSAGQUNP-UHFFFAOYSA-N
Synonym 4-methylamino benzenesulfonic acid, 4-methylamino benzenesulphonic acid, n-methylsulphanilic acid, n-methylsulfanilic acid, benzenesulfonic acid,4-methylamino, n-methylaniline-p-sulfonic acid, maybridge1_004198, n-methylsulfanilic acid, 98+%
PubChem CID 90505
IUPAC Name 4-(methylamino)benzenesulfonic acid
SMILES CNC1=CC=C(C=C1)S(=O)(=O)O

Specifications

Melting Point ∽260°C (decomposition)
Quantity 25 g
Beilstein 2094129
Formula Weight 187.21
Percent Purity ≥98%
Chemical Name or Material N-Methylsulfanilic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-256-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.