Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methyltyramine hydrochloride | 13062-76-5 | MFCD01707290 | 99.4% | 187.67 g/mol | C9H14ClNO | 5 G
SDP

Catalog No. 5000280828
Encompass_Preferred

N-methyltyramine hydrochloride is the hydrochloride salt of N-methyltyramine, a naturally occurring phenethylamine isolated from Citrus species. Supplied as a white to off-white solid for research use, it is employed as a biochemical and pharmacological reference compound, including studies of adrenergic activity.

  • High purity (99.4%).
  • Molecular formula C9H14ClNO; molecular weight 187.67 g/mol.
  • CAS number 13062-76-5 for precise identification.
  • White to off-white solid suitable for analytical applications.
  • Available in small lab pack sizes, including 5 G.
  • Store sealed, away from moisture; short-term storage in solution at -20°C to -80°C.

Catalog No. 50-002-80828 Supplier Medchemexpress LLC Supplier No. HYW1531595G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-methyltyramine hydrochloride is the hydrochloride salt of N-methyltyramine, a naturally occurring phenethylamine isolated from Citrus species. Supplied as a white to off-white solid for research use, it is employed as a biochemical and pharmacological reference compound, including studies of adrenergic activity.

  • High purity (99.4%).
  • Molecular formula C9H14ClNO; molecular weight 187.67 g/mol.
  • CAS number 13062-76-5 for precise identification.
  • White to off-white solid suitable for analytical applications.
  • Available in small lab pack sizes, including 5 G.
  • Store sealed, away from moisture; short-term storage in solution at -20°C to -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.