Learn More
Cusabio Technology LLC N-myc proto-oncogene protein
Catalog No. 5000050647
Encompass_Preferred
This kit detects MYCN in human serum plasma tissue homogenates and cell lysates with high precision It employs a quantitative sandwich ELISA method offering a detection range of 15 6 pg/mL to 1000 pg/mL and a sensitivity of 3 9 pg/mL The assay requires a sample volume of 50-100 L and an incubation time of 1-5 hours with absorbance measured at 450 nm Designed for reliable performance it enables accurate measurement of MYCN protein levels in diverse biological matrices supporting research applications such as cancer studies biomarker analysis and cellular pathway investigations The kit s standardized protocol ensures reproducibility and ease of use in both laboratory and clinical settings
Catalog No. 50-000-50647 Supplier Cusabio Technology LLC Supplier No. CSBEL015278HU
This kit detects MYCN in human serum plasma tissue homogenates and cell lysates with high precision It employs a quantitative sandwich ELISA method offering a detection range of 15 6 pg/mL to 1000 pg/mL and a sensitivity of 3 9 pg/mL The assay requires a sample volume of 50-100 L and an incubation time of 1-5 hours with absorbance measured at 450 nm Designed for reliable performance it enables accurate measurement of MYCN protein levels in diverse biological matrices supporting research applications such as cancer studies biomarker analysis and cellular pathway investigations The kit s standardized protocol ensures reproducibility and ease of use in both laboratory and clinical settings