Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-myristoyl-D-erythro-sphingosine | 123408-74-2 | ≥99.0% | 509.85 | C32H63NO3 | 5MG
SDP

Catalog No. 5000237796
Encompass_Preferred

C14 Ceramide (d18:1/14:0), chemical name N-myristoyl-D-erythro-sphingosine (CAS 123408-74-2), is an endogenous N-acyl sphingosine produced by ceramide synthetase 6 and used in research on Parkinson's disease and diabetes. The compound has formula C32H63NO3, molecular weight 509.85, and is supplied in small research pack sizes for laboratory use.

  • Endogenous ceramide produced by ceramide synthetase 6.
  • Used for Parkinson's disease and diabetes research.
  • High purity: ≥99.0%.
  • Chemical formula C32H63NO3 and molecular weight 509.85.
  • Available in small research pack sizes, including 5 mg.
  • Storage: powder -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-37796 Supplier Medchemexpress LLC Supplier No. HY124177A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C14 Ceramide (d18:1/14:0), chemical name N-myristoyl-D-erythro-sphingosine (CAS 123408-74-2), is an endogenous N-acyl sphingosine produced by ceramide synthetase 6 and used in research on Parkinson's disease and diabetes. The compound has formula C32H63NO3, molecular weight 509.85, and is supplied in small research pack sizes for laboratory use.

  • Endogenous ceramide produced by ceramide synthetase 6.
  • Used for Parkinson's disease and diabetes research.
  • High purity: ≥99.0%.
  • Chemical formula C32H63NO3 and molecular weight 509.85.
  • Available in small research pack sizes, including 5 mg.
  • Storage: powder -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.