Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N′-1,3-phenylenebis-[2,3-dihydroxy-benzamide] | 368449-04-1 | 98.6% | 10 MG
SDP

Catalog No. 5000232924
Encompass_Preferred

MST-312 (telomerase inhibitor IX) is a small-molecule telomerase inhibitor used in research to study telomerase-dependent pathways and anticancer activity. It is a bis-catechol derivative of epigallocatechin gallate and is used in cell-based assays to reduce proliferation and induce apoptosis in sensitive lines.

  • Inhibits telomerase activity in cell-based assays.
  • Derivative of epigallocatechin gallate (EGCG).
  • High purity (≈98.6%).
  • Molecular weight 380.35 g/mol.
  • Available as solid and as 10 mM solution in DMSO.
  • Recommended storage: powder at -20°C; in solvent at -80°C.

Catalog No. 50-002-32924 Supplier Medchemexpress LLC Supplier No. HY12014510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MST-312 (telomerase inhibitor IX) is a small-molecule telomerase inhibitor used in research to study telomerase-dependent pathways and anticancer activity. It is a bis-catechol derivative of epigallocatechin gallate and is used in cell-based assays to reduce proliferation and induce apoptosis in sensitive lines.

  • Inhibits telomerase activity in cell-based assays.
  • Derivative of epigallocatechin gallate (EGCG).
  • High purity (≈98.6%).
  • Molecular weight 380.35 g/mol.
  • Available as solid and as 10 mM solution in DMSO.
  • Recommended storage: powder at -20°C; in solvent at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.