Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N,2-trimethyl-5-((4-(2-(4-(3-methylureido)phenyl)pyridin-4-yl)pyrimidin-2-yl)amino)benzenesulfonam | 1402452-10-1 | 99.9% | 517.60 | C26H27N7O3S | 50MG
SDP

Catalog No. 5000237011
Encompass_Preferred

hSMG-1 inhibitor 11e is a selective small-molecule inhibitor of the human SMG-1 kinase provided as a solid research reagent. It is intended for use in biochemical and cellular assays that probe PI3K-related protein kinase activity. The compound has a high reported purity and established solubility in DMSO, with recommended sealed storage away from moisture and light.

  • Molecular formula: C26H27N7O3S and molecular weight approximately 517.60.
  • High reported purity suitable for research applications.
  • Physical form: solid; appearance white to light yellow.
  • Solubility: readily soluble in DMSO (about 50 mg/mL); ultrasonic agitation may be required.
  • Storage: keep sealed, away from moisture and light; in solvent store at -80°C for long term.
  • Intended use: in vitro kinase inhibition and mechanistic studies.

Catalog No. 50-002-37011 Supplier Medchemexpress LLC Supplier No. HY12476050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

hSMG-1 inhibitor 11e is a selective small-molecule inhibitor of the human SMG-1 kinase provided as a solid research reagent. It is intended for use in biochemical and cellular assays that probe PI3K-related protein kinase activity. The compound has a high reported purity and established solubility in DMSO, with recommended sealed storage away from moisture and light.

  • Molecular formula: C26H27N7O3S and molecular weight approximately 517.60.
  • High reported purity suitable for research applications.
  • Physical form: solid; appearance white to light yellow.
  • Solubility: readily soluble in DMSO (about 50 mg/mL); ultrasonic agitation may be required.
  • Storage: keep sealed, away from moisture and light; in solvent store at -80°C for long term.
  • Intended use: in vitro kinase inhibition and mechanistic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.