Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,2-Trimethyl-5-nitrobenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. T3112200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3112200MG 200 mg
T31121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3112200MG Supplier TCI America Supplier No. T3112200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 433695-36-4
Molecular Formula C9H12N2O4S
Molecular Weight (g/mol) 244.265
MDL Number MFCD03039912
InChI Key IFIUFCJFLGCQPH-UHFFFAOYSA-N
Synonym unii-03g869pr3p, benzenesulfonamide, n,n,2-trimethyl-5-nitro, 5-nitro-2,n,n-trimethylbenzenesulfonamide, d00mbg, cambridge id 6971444, n,n,2-trimethyl-5-nitro-benzenesulfonamide, brl hplc , solid, n,n,2-trimethyl-5-nitrobenzene-1-sulfonamide, 2-methyl-5-nitro n,n-dimethyl benzenesulfonamide, dimethyl 2-methyl-5-nitrophenyl sulfonyl amine
PubChem CID 2921148
IUPAC Name N,N,2-trimethyl-5-nitrobenzenesulfonamide
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])S(=O)(=O)N(C)C

Specifications

Melting Point 73°C
Color Yellow
Quantity 200 mg
Formula Weight 244.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,2-Trimethyl-5-nitrobenzenesulfonamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.