Learn More
Medchemexpress LLC N,N,6-triethyl-9,10-didehydro-1-(1-oxopropyl)-ergoline-8β-carboxamide | 2648317-95-5 | 99.5% | C24H28N2O3 | 10MG
SDP
Catalog No. 5000260219
Encompass_Preferred
GLP-1R modulator L7-028 is a small-molecule positive allosteric modulator of the glucagon-like peptide-1 receptor (GLP-1R) that enhances GLP-1 binding via a transmembrane site. Supplied as a solid or as a 10 mM solution in DMSO for research use, it demonstrates activity in the low micromolar range and is provided with high purity.
  • Enhances GLP-1 binding to GLP-1R via a transmembrane site (EC50 11.01 ± 2.73 μM).
  • High purity, approx. 99.5%.
  • Available as solid and as 10 mM solution in DMSO.
  • Soluble in DMSO at 100 mg/mL; ultrasonic recommended for dissolution.
  • Stable under recommended storage: powder -20°C for long term; solution -80°C for extended storage.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-60219 Supplier Medchemexpress LLC Supplier No. HY14184210MG
GLP-1R modulator L7-028 is a small-molecule positive allosteric modulator of the glucagon-like peptide-1 receptor (GLP-1R) that enhances GLP-1 binding via a transmembrane site. Supplied as a solid or as a 10 mM solution in DMSO for research use, it demonstrates activity in the low micromolar range and is provided with high purity.
  • Enhances GLP-1 binding to GLP-1R via a transmembrane site (EC50 11.01 ± 2.73 μM).
  • High purity, approx. 99.5%.
  • Available as solid and as 10 mM solution in DMSO.
  • Soluble in DMSO at 100 mg/mL; ultrasonic recommended for dissolution.
  • Stable under recommended storage: powder -20°C for long term; solution -80°C for extended storage.
  • Intended for research use only; not for human or veterinary use.