Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(2,2-diethoxyethyl)methylamine 97%, Thermo Scientific™

Catalog No. AC152001000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC152001000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC152001000 Supplier Thermo Scientific Chemicals Supplier No. 152001000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6948-86-3
Molecular Formula C13H29NO4
Molecular Weight (g/mol) 263.378
InChI Key DPLOXPRJCFWURN-UHFFFAOYSA-N
Synonym n,n-bis 2,2-diethoxyethyl methylamine, n-2,2-diethoxyethyl-2,2-diethoxy-n-methylethanamine, 2,2'-methyliminobis acetaldehyde diethyl acetal, n-2,2-bis ethyloxy ethyl-2,2-bis ethyloxy-n-methylethanamine, dploxprjcfwurn-uhfffaoysa, n,n-bis 2,2-diethoxyethyl methyl amine, bis 2,2-diethoxyethyl methyl amine, ethanamine, n-2,2-diethoxyethyl-2,2-diethoxy-n-methyl, n-2,2-diethoxyethyl-2,2-diethoxy-n-methylethanamine #, 2,2 inverted exclamation marka-methyliminobis acetaldehyde diethyl acetal
PubChem CID 81381
IUPAC Name N-(2,2-diethoxyethyl)-2,2-diethoxy-N-methylethanamine
SMILES CCOC(CN(C)CC(OCC)OCC)OCC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Linear Formula [(C2H5O)2CHCH2]2NCH3
Quantity 100 g
Formula Weight 263.38
Chemical Name or Material N, N-Bis(2, 2-diethoxyethyl)methylamine, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.