Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(2-aminobenzal)ethylenediamine 97.0+%, TCI America™
SDP

Catalog No. A02611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A02611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A02611G Supplier TCI America Supplier No. A02611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4408-47-3
Molecular Formula C16H18N4
Molecular Weight (g/mol) 266.35
MDL Number MFCD00059854
InChI Key ZFIFWHZGMOGXDV-UHFFFAOYSA-N
Synonym 2-Aminobenzaldehyde-ethylene-diimine, Ethylenedinitrilobis(o-toluidine)
PubChem CID 6432140
IUPAC Name 2-{[(2-{[(2-aminophenyl)methylidene]amino}ethyl)imino]methyl}aniline
SMILES NC1=CC=CC=C1C=NCCN=CC1=CC=CC=C1N

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 1 g
Formula Weight 266.35
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(2-aminobenzal)ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.