Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N'-Bis(2-aminoethyl)-1,3-propanediamine

Catalog No. 3331311G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-313-11G 1 g
33-313-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-313-11G Supplier Sigma Aldrich Supplier No. 3331311G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,nα-bis(2-aminoethyl)-1,3-propanediamine is a high affinity cu(ii) chelator
  • It inhibits mitochondrial cytochrome c oxidase by causing cellular cu deficiency in a human promyelocytic leukemia cell line (hl-60).n,nα-bis(2-aminoethyl)-1,3-propanediamine was used to study the regulation of gene that encodes the periplasmic superoxide dismutase of escherichia coli in response to copper

Specifications

CAS 4741-99-5
Density 0.96 g/mL (at 25°C (literature))
Boiling Point 142°C to 145°C (8 mmHg)
Molecular Formula C7H20N4
Refractive Index n20/D 1.494 (literature)
Linear Formula H2NCH2CH2NH(CH2)3 NHCH2CH2NH2
MDL Number MFCD00008174
Quantity 1 g
Molecular Weight (g/mol) 160.26
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.