Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(2-aminoethyl)-1,3-propanediamine, 97%, Thermo Scientific™

Catalog No. AC402260250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC402260250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402260250 Supplier Thermo Scientific Chemicals Supplier No. 402260250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4741-99-5
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H20N4
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Linear Formula NH2(CH2)2NH(CH2)3NH(CH2)2NH2
Quantity 25g
Synonym n,n'-bis 2-aminoethyl-1,3-propanediamine, 1,4,8,11-tetraazaundecane, 2,3,2-tetramine, n,n'-bis 2-aminoethyl propane-1,3-diamine, 1,4,8,11-tetrazaundecane, ethylenetrimethyleneethylenetetramine, 1,3-propanediamine, n,n'-bis 2-aminoethyl, 1,9-diamino-3,7-diazanonane, n1,n1'-propane-1,3-diyl bis ethane-1,2-diamine, 3,7-diazanonane-1,9-diamine
InChI Key UWMHHZFHBCYGCV-UHFFFAOYSA-N
SMILES C(CNCCN)CNCCN
IUPAC Name N,N'-bis(2-aminoethyl)propane-1,3-diamine
Molecular Weight (g/mol) 160.265
PubChem CID 78479
ChEBI CHEBI:30348
Formula Weight 160.26
Percent Purity ≥96.0%
Chemical Name or Material N, N'-Bis(2-aminoethyl)-1, 3-propanediamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-254-1
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.