Learn More
Medchemexpress LLC N, N'-bis(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid | 35369-53-0 | ≥95.0% | 461.34 | C20H26Cl2N2O6 | 100 MG
SDP
Catalog No. 5000299315
Encompass_Preferred
HBED dihydrochloride is the dihydrochloride salt of N,N'-bis(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid, an orally active hexadentate phenolic aminocarboxylate that chelates iron. It is provided for biochemical research and assay use, supplied as a solid and as ready-to-use DMSO solutions, and is commonly used in iron chelation studies and related biochemical applications.
  • Hexadentate phenolic aminocarboxylate iron chelator.
  • Available as solid and DMSO solution formats for flexibility.
  • High solubility in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Documented purity for research use.
  • Used for iron chelation and biochemical assay applications.
Catalog No. 50-002-99315 Supplier Medchemexpress LLC Supplier No. HYW035520100MG
HBED dihydrochloride is the dihydrochloride salt of N,N'-bis(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid, an orally active hexadentate phenolic aminocarboxylate that chelates iron. It is provided for biochemical research and assay use, supplied as a solid and as ready-to-use DMSO solutions, and is commonly used in iron chelation studies and related biochemical applications.
  • Hexadentate phenolic aminocarboxylate iron chelator.
  • Available as solid and DMSO solution formats for flexibility.
  • High solubility in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Documented purity for research use.
  • Used for iron chelation and biochemical assay applications.