Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N'-bis(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid dihydrochloride | 35369-53-0 | MFCD04113603 | >95.0% | 461.34 | C20H26Cl2N2O6 | 1 ML
SDP

Catalog No. 5000299847
Encompass_Preferred

HBED dihydrochloride is an orally active, hexadentate phenolic aminocarboxylate iron chelator (N,N'-bis(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid dihydrochloride) supplied for research use in solution and solid formats for studies of iron binding and chelation biology.

  • Orally active hexadentate iron chelator.
  • Suitable for iron chelation research and biochemical assays.
  • Supplied as ready-to-use 10 mM in DMSO solution and as solid material.
  • Solid sizes available: 100 mg, 250 mg, 500 mg, 1 g.
  • Molecular weight 461.34; formula C20H26Cl2N2O6.
  • Typical purity specification ≥95.0% (batch dependent).

Catalog No. 50-002-99847 Supplier Medchemexpress LLC Supplier No. HYW03552010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HBED dihydrochloride is an orally active, hexadentate phenolic aminocarboxylate iron chelator (N,N'-bis(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid dihydrochloride) supplied for research use in solution and solid formats for studies of iron binding and chelation biology.

  • Orally active hexadentate iron chelator.
  • Suitable for iron chelation research and biochemical assays.
  • Supplied as ready-to-use 10 mM in DMSO solution and as solid material.
  • Solid sizes available: 100 mg, 250 mg, 500 mg, 1 g.
  • Molecular weight 461.34; formula C20H26Cl2N2O6.
  • Typical purity specification ≥95.0% (batch dependent).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.