Learn More
Medchemexpress LLC N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid | 10191-18-1 | 98.0% | 500 G
SDP
Catalog No. 5000410619
Encompass_Preferred
BES is a secondary standard biochemical buffer used in diagnostic testing. It offers a stable pH environment for various applications.
  • Effective pH range of 6.4 to 7.8
  • pKa of 7.15 at 20 °C
  • Suitable for diagnostic testing areas
Catalog No. 50-004-10619 Supplier Medchemexpress LLC Supplier No. HYD0867500G
BES is a secondary standard biochemical buffer used in diagnostic testing. It offers a stable pH environment for various applications.
  • Effective pH range of 6.4 to 7.8
  • pKa of 7.15 at 20 °C
  • Suitable for diagnostic testing areas