Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N'-Bis(2-hydroxyethyl)ethylenediamine

Catalog No. 26887950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-887-950G 50 g
26-887-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-887-950G Supplier Sigma Aldrich Supplier No. 26887950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N'-Bis(2-hydroxyethyl)ethylenediamine

Specifications

CAS 4439-20-7
Melting Point 98°C to 100°C (lit.)
Molecular Formula C6H16N2O2
Linear Formula HOCH2CH2NHCH2CH2NHCH2CH2OH
MDL Number MFCD00002842
Quantity 50 g
Synonym 2,2 -(Ethylenediimino)diethanol
Molecular Weight (g/mol) 148.2
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.