Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(2-hydroxyethyl)ethylenediamine 98.0+%, TCI America™
SDP

Catalog No. B380725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B380725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B380725G Supplier TCI America Supplier No. B380725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An amino alcohol building block for chemical research.

Specifications

CAS 4439-20-7
Melting Point 99°C
Color White-Yellow
Molecular Formula C6H16N2O2
Quantity 25 g
MDL Number MFCD00002842
UN Number 3259
Synonym n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol
InChI Key GFIWSSUBVYLTRF-UHFFFAOYSA-N
SMILES C(CNCCO)NCCO
IUPAC Name 2-[2-(2-hydroxyethylamino)ethylamino]ethanol
Molecular Weight (g/mol) 148.206
PubChem CID 78179
Formula Weight 148.21
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(2-hydroxyethyl)ethylenediamine
Show More Show Less
EINECSNumber (2)-2409
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.