Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(2-hydroxyethyl)-p-toluenesulfonamide 97.0+%, TCI America™
SDP

Catalog No. B226325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B226325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B226325G Supplier TCI America Supplier No. B226325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7146-67-0
Molecular Formula C11H17NO4S
Molecular Weight (g/mol) 259.32
MDL Number MFCD00020582
InChI Key NHFJDXPINIIUQG-UHFFFAOYSA-N
Synonym n,n-bis 2-hydroxyethyl-4-methylbenzenesulfonamide, n,n-bis 2-hydroxyethyl-p-toluenesulfonamide, n,n-bis-2-hydroxyethyl-p-toluenesulfonamide, benzenesulfonamide, n,n-bis 2-hydroxyethyl-4-methyl, n,n-bis 2-hydroxyethyl-p-toluenesulphonamide, benzenesulfonamide,n-bis 2-hydroxyethyl-4-methyl, bis 2-hydroxyethyl 4-methylphenyl sulfonyl amine, n-tosyldiethanolamine, acmc-209oio, n,n-bis 2-hydroxyethyl-4-toluenesulfonamide
PubChem CID 81565
IUPAC Name N,N-bis(2-hydroxyethyl)-4-methylbenzene-1-sulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(CCO)CCO

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 25 g
Formula Weight 259.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(2-hydroxyethyl)-p-toluenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.