Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(2-hydroxypropyl)-N'-(α,α-dimethyl-3-isopropenylbenzyl)urea, 98%, Thermo Scientific™

Catalog No. AC402560250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC402560250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402560250 Supplier Thermo Scientific Chemicals Supplier No. 402560250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 214531-79-0
CAS Min % 97.5
CAS Max % 100.0
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C19H30N2O3
Quantity 25g
Synonym 1,1-bis 2-hydroxypropyl-3-2-3-prop-1-en-2-yl phenyl propan-2-yl urea, n,n-bis 2-hydroxypropyl-n'-alpha,alpha-dimethyl-3-isopropenylbenzyl urea, 3,3-bis 2-hydroxypropyl-1-2-3-prop-1-en-2-yl phenyl propan-2-yl urea, 1,1-bis 2-hydroxypropyl-3-2-3-prop-1-en-2-ylphenyl propan-2-yl urea, n,n-bis 2-hydroxypropyl-n'-a,a-dimethyl-3-isopropenylbenzyl urea, n,n-bis 2-hydroxypropyl 1-methyl-1-3-1-methylvinyl phenyl ethyl amino carb oxamide
InChI Key CNWWIASFGWOBBQ-UHFFFAOYSA-N
SMILES CC(CN(CC(C)O)C(=O)NC(C)(C)C1=CC=CC(=C1)C(=C)C)O
IUPAC Name 1,1-bis(2-hydroxypropyl)-3-[2-(3-prop-1-en-2-ylphenyl)propan-2-yl]urea
Molecular Weight (g/mol) 334.46
PubChem CID 2734977
Formula Weight 334.45
Chemical Name or Material N, N-Bis(2-hydroxypropyl)-N'-(α, α-dimethyl-3-isopropenylbenzyl)urea
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.