Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N'-bis(3,4-dichlorophenyl)urea | 4300-43-0 | MFCD00171464 | 95.0% | 350.03 | C13H8Cl4N2O | 250MG
SDP

Catalog No. 5000238408
Encompass_Preferred

MMV665852 is a research antischistosomal small molecule that inhibits worm viability in vitro and reduces worm burden in mice infected with Schistosoma mansoni. It is chemically N,N'-bis(3,4-dichlorophenyl)urea (CAS 4300-43-0), with molecular formula C13H8Cl4N2O and molecular weight 350.03 g/mol. Product documentation and the certificate of analysis should be consulted for purity and storage instructions.

  • Inhibits worm viability in vitro.
  • Reduces worm burden in Schistosoma mansoni-infected mice.
  • Chemical name N,N'-bis(3,4-dichlorophenyl)urea; CAS 4300-43-0.
  • Molecular formula C13H8Cl4N2O and molecular weight 350.03 g/mol.
  • Available in multiple packaging sizes (100 mg-10 g).
  • Purity and QC data provided in the certificate of analysis.

Catalog No. 50-002-38408 Supplier Medchemexpress LLC Supplier No. HY124018250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MMV665852 is a research antischistosomal small molecule that inhibits worm viability in vitro and reduces worm burden in mice infected with Schistosoma mansoni. It is chemically N,N'-bis(3,4-dichlorophenyl)urea (CAS 4300-43-0), with molecular formula C13H8Cl4N2O and molecular weight 350.03 g/mol. Product documentation and the certificate of analysis should be consulted for purity and storage instructions.

  • Inhibits worm viability in vitro.
  • Reduces worm burden in Schistosoma mansoni-infected mice.
  • Chemical name N,N'-bis(3,4-dichlorophenyl)urea; CAS 4300-43-0.
  • Molecular formula C13H8Cl4N2O and molecular weight 350.03 g/mol.
  • Available in multiple packaging sizes (100 mg-10 g).
  • Purity and QC data provided in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.