Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis[3-(tert-butoxycarbonylamino)propyl]glycinamide 93.0+%, TCI America™
SDP

Catalog No. B2419200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2419200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2419200MG Supplier TCI America Supplier No. B2419200MG

Chemical Identifiers

CAS 1822851-77-3
Molecular Formula C18H36N4O5
Molecular Weight (g/mol) 388.509
InChI Key LUUNBXYJZAFAKK-UHFFFAOYSA-N
Synonym N,N′-Bis[3-(Boc-amino)propyl]glycinamide
PubChem CID 44629798
IUPAC Name tert-butyl N-[3-[[2-[3-[(2-methylpropan-2-yl)oxycarbonylamino]propylamino]acetyl]amino]propyl]carbamate
SMILES CC(C)(C)OC(=O)NCCCNCC(=O)NCCCNC(=O)OC(C)(C)C

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 388.51
Percent Purity ≥93.0% (T)
Physical Form Liquid
Chemical Name or Material N,N′-Bis[3-(tert-butoxycarbonylamino)propyl]glycinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.