Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine 98.0+%, TCI America™
SDP

Catalog No. B45505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45505G 5 g
B455025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45505G Supplier TCI America Supplier No. B45505G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 499128-71-1
Molecular Formula C30H22BrN
Molecular Weight (g/mol) 476.417
MDL Number MFCD20261458
InChI Key BTFIECQCKYNJTN-UHFFFAOYSA-N
PubChem CID 11454354
IUPAC Name N-(4-bromophenyl)-4-phenyl-N-(4-phenylphenyl)aniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N(C3=CC=C(C=C3)C4=CC=CC=C4)C5=CC=C(C=C5)Br

Specifications

Melting Point 171°C
Color White
Quantity 5 g
Synonym 4-Bromo-4′C,4′C′C-diphenyltriphenylamine
Formula Weight 476.42
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.