Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(4-bromophenyl)-2,4,6-trimethylaniline 98.0+%, TCI America™
SDP

Catalog No. B55075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55075G 5 g
B55071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55075G Supplier TCI America Supplier No. B55075G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 663943-27-9
Molecular Formula C21H19Br2N
Molecular Weight (g/mol) 445.198
InChI Key XZCBYXUKPMELOQ-UHFFFAOYSA-N
PubChem CID 22930861
IUPAC Name N,N-bis(4-bromophenyl)-2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1)C)N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)C

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 5 g
Formula Weight 445.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(4-bromophenyl)-2,4,6-trimethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.