Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N'-Bis(acryloyl)cystamine, Electrophoresis Reagent, 98%

Click to view available options
Quantity:
1g
5g
25g
60984-57-8
C10H16N2O2S2
260.37
MFCD00036225
DJVKJGIZQFBFGS-UHFFFAOYSA-N
n,n'-bis acryloyl cystamine,n,n'-bisacrylylcystamine,2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis,2,2'-bisacrylamino diethyldisulfide,bis-acrylylcystamine: bac,n,n'-dithiodi-2,1-ethanediyl bis acrylamide,n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide,n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide,acmc-20ak9p,n,n-bis-acrylylcystamine
100602
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
C=CC(=O)NCCSSCCNC(=O)C=C
60984-57-8
C10H16N2O2S2
260.37
MFCD00036225
DJVKJGIZQFBFGS-UHFFFAOYSA-N
n,n'-bis acryloyl cystamine,n,n'-bisacrylylcystamine,2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis,2,2'-bisacrylamino diethyldisulfide,bis-acrylylcystamine: bac,n,n'-dithiodi-2,1-ethanediyl bis acrylamide,n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide,n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide,acmc-20ak9p,n,n-bis-acrylylcystamine
100602
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
C=CC(=O)NCCSSCCNC(=O)C=C
Applications
BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.

Solubility
Soluble at 50 mg/ml in acetic acid

Notes
Store at -20°C. Incompatible with heat, oxidizing agents.
Color White
Melting Point 123°C to 124°C
Quantity 1g
Solubility Information Soluble at 50mg/mL in acetic acid
Formula Weight 260.37
Physical Form Powder
Percent Purity 98%
Chemical Name or Material N,N'-Bis(acryloyl)cystamine, Electrophoresis Reagent

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : −30°C to -10°C