Learn More
Medchemexpress LLC Nα,Nα-bis(carboxymethyl)-L-lysine | 113231-05-3 | 98.8% | 262.26 | C10H18N2O6 | 100 G
SDP
Catalog No. 5000215486
Encompass_Preferred
Nα,Nα-Bis(carboxymethyl)-L-lysine is a research-grade amino acid derivative and potent competitive inhibitor of the bitter taste receptor TAS2R4 (IC50 = 59 nM). It is used in receptor pharmacology, structure-activity relationship studies, and biochemical assays requiring a selective TAS2R4 antagonist.
  • Competitive inhibitor of bitter taste receptor 4 (IC50 = 59 nM).
  • High purity suitable for biochemical and pharmacological assays (98.8%).
  • Solid reagent compatible with standard solvents used in receptor studies.
  • Molecular weight 262.26; molecular formula C10H18N2O6.
  • Available in lab-scale quantities (for example, 100 g) for screening and validation.
Catalog No. 50-002-15486 Supplier Medchemexpress LLC Supplier No. HY100047100G
Nα,Nα-Bis(carboxymethyl)-L-lysine is a research-grade amino acid derivative and potent competitive inhibitor of the bitter taste receptor TAS2R4 (IC50 = 59 nM). It is used in receptor pharmacology, structure-activity relationship studies, and biochemical assays requiring a selective TAS2R4 antagonist.
  • Competitive inhibitor of bitter taste receptor 4 (IC50 = 59 nM).
  • High purity suitable for biochemical and pharmacological assays (98.8%).
  • Solid reagent compatible with standard solvents used in receptor studies.
  • Molecular weight 262.26; molecular formula C10H18N2O6.
  • Available in lab-scale quantities (for example, 100 g) for screening and validation.