Learn More
Medchemexpress LLC Nα,Nα-bis(carboxymethyl)-L-lysine | 113231-05-3 | MFCD00237437 | 98.8% | 262.26 | C10H18N2O6 | 10 G
SDP
Catalog No. 5000215209
Encompass_Preferred
Nα,Nα-Bis(carboxymethyl)-L-lysine is a research-grade small molecule that competitively inhibits the bitter taste receptor T2R4, with a reported IC50 of approximately 59 nM. It is used in receptor pharmacology and cell-based assays to probe T2R4-mediated signaling; in vitro studies show reduction of basal receptor activity, while limited in vivo effects on bitter taste behavior have been reported.
  • Competitive inhibitor of bitter taste receptor 4 (IC50 ≈59 nM).
  • Suitable for receptor pharmacology and cell-based assays.
  • Shown in vitro to reduce basal activity of constitutively active T2R4 mutants.
  • Reported minimal in vivo effect on bitter taste licking behavior at 10 μM.
  • High purity material (98.8%) for research use.
  • Molecular weight 262.26; formula C10H18N2O6.
Catalog No. 50-002-15209 Supplier Medchemexpress LLC Supplier No. HY10004710G
Nα,Nα-Bis(carboxymethyl)-L-lysine is a research-grade small molecule that competitively inhibits the bitter taste receptor T2R4, with a reported IC50 of approximately 59 nM. It is used in receptor pharmacology and cell-based assays to probe T2R4-mediated signaling; in vitro studies show reduction of basal receptor activity, while limited in vivo effects on bitter taste behavior have been reported.
  • Competitive inhibitor of bitter taste receptor 4 (IC50 ≈59 nM).
  • Suitable for receptor pharmacology and cell-based assays.
  • Shown in vitro to reduce basal activity of constitutively active T2R4 mutants.
  • Reported minimal in vivo effect on bitter taste licking behavior at 10 μM.
  • High purity material (98.8%) for research use.
  • Molecular weight 262.26; formula C10H18N2O6.