Learn More
Medchemexpress LLC Nα,Nα-bis(carboxymethyl)-L-lysine | 113231-05-3 | MFCD00237437 | 98.8% | 262.26 | C10H18N2O6 | 25 G
SDP
Catalog No. 5000215304
Encompass_Preferred
Nα,Nα-bis(carboxymethyl)-L-lysine is an amino-acid derivative provided as a research-grade reagent for cellular and receptor pharmacology studies. It functions as a competitive inhibitor of the bitter taste receptor T2R4 (IC50 = 59 nM) and is supplied as a white to off-white powder for laboratory assays.
  • Competitive T2R4 inhibitor (IC50 = 59 nM).
  • High purity, 98.8% by supplier specification.
  • Molecular formula C10H18N2O6; molecular weight 262.26.
  • Available as a white to off-white powder for laboratory use.
  • Decreases basal activity of a constitutively active T2R4 mutant in vitro.
  • Suitable for cell-based assays investigating bitter receptor signaling.
Catalog No. 50-002-15304 Supplier Medchemexpress LLC Supplier No. HY10004725G
Nα,Nα-bis(carboxymethyl)-L-lysine is an amino-acid derivative provided as a research-grade reagent for cellular and receptor pharmacology studies. It functions as a competitive inhibitor of the bitter taste receptor T2R4 (IC50 = 59 nM) and is supplied as a white to off-white powder for laboratory assays.
  • Competitive T2R4 inhibitor (IC50 = 59 nM).
  • High purity, 98.8% by supplier specification.
  • Molecular formula C10H18N2O6; molecular weight 262.26.
  • Available as a white to off-white powder for laboratory use.
  • Decreases basal activity of a constitutively active T2R4 mutant in vitro.
  • Suitable for cell-based assays investigating bitter receptor signaling.