Learn More
Medchemexpress LLC Nα,Nα-bis(carboxymethyl)-L-lysine | 113231-05-3 | MFCD00237437 | 98.8% | 262.26 g/mol | C10H18N2O6 | 5 G
SDP
Catalog No. 5000215491
Encompass_Preferred
Nα,Nα-bis(carboxymethyl)-L-lysine is a small-molecule competitive inhibitor of the bitter taste receptor TAS2R4, supplied for receptor pharmacology and bitter-receptor research. The compound has a reported IC50 of 59 nM against TAS2R4 and is provided with analytical purity documentation for use in biochemical assays.
  • Competitive inhibitor of bitter taste receptor 4 (IC50 59 nM).
  • High purity suitable for biochemical and pharmacology assays.
  • Available in multiple pack sizes including 5 G.
  • Molecular weight 262.26 g/mol; formula C10H18N2O6.
  • Includes RP-HPLC documentation for purity verification.
Catalog No. 50-002-15491 Supplier Medchemexpress LLC Supplier No. HY1000475G
Nα,Nα-bis(carboxymethyl)-L-lysine is a small-molecule competitive inhibitor of the bitter taste receptor TAS2R4, supplied for receptor pharmacology and bitter-receptor research. The compound has a reported IC50 of 59 nM against TAS2R4 and is provided with analytical purity documentation for use in biochemical assays.
  • Competitive inhibitor of bitter taste receptor 4 (IC50 59 nM).
  • High purity suitable for biochemical and pharmacology assays.
  • Available in multiple pack sizes including 5 G.
  • Molecular weight 262.26 g/mol; formula C10H18N2O6.
  • Includes RP-HPLC documentation for purity verification.