Learn More
Cayman Chemical N N-bIs-CbzAlethIn 10mg
SDP
Catalog No. 502980663
Encompass_Preferred
An active ester used in the synthesis of β-alethine
Catalog No. 50-298-0663 Supplier Cayman Chemical Supplier No. 2087810 MG
An active ester used in the synthesis of β-alethine