Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(β-hydroxyethyl)aniline, MP Biomedicals

Catalog No. ICN217370 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217370 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217370 Supplier MP Biomedicals, Inc Supplier No. 0521737080

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-07-0
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD00002845
InChI Key OJPDDQSCZGTACX-UHFFFAOYSA-N
Synonym n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
PubChem CID 8416
IUPAC Name 2-[(2-hydroxyethyl)(phenyl)amino]ethan-1-ol
SMILES OCCN(CCO)C1=CC=CC=C1

Specifications

Quantity 100 g
Formula Weight 181.24
Chemical Name or Material N, N-Bis(β-Hydroxyethyl)Aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.