Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Bis(phosphonomethyl)glycine

Catalog No. 1514950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-149-50G 50 g
15-149-10G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-149-50G Supplier Sigma Aldrich Supplier No. 1514950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-bis(phosphonomethyl)glycine was present in the ligand bound to human apotransferrin.application: complexing agent; pk-values: pk1: -+2, pk2: 2.0, pk3: 5.20, pk4: 6.77, pk5: 10.89 anti-diabetic; delay onset of diabetes in non-obese nod miceglyphosine binds to a pocket of mhc class ii molecules i-ag7 and dq8, and modify t cell responses to the autoantigenic insulin b chain fragment 9-23 (b:9-23) peptide
  • Glyphosine stimulate il-10 production and delay onset of diabetes in non-obese nod mice. anti-diabetic; delay onset of diabetes in non-obese nod mice

Specifications

CAS 2439-99-8
Molecular Formula C4H11NO8P2
Linear Formula C4H11NO8P2
MDL Number MFCD00056670
Quantity 50 g
Synonym N-(Carboxymethyl)iminodi(methylphosphinic acid); Glyphosine
Molecular Weight (g/mol) 263.08
Percent Purity ≥98.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.