Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(salicylidene)-1,2-propanediamine 98.0+%, TCI America™
SDP

Catalog No. D130725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D130725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D130725G Supplier TCI America Supplier No. D130725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94-91-7
Molecular Formula C17H18N2O2
Molecular Weight (g/mol) 282.343
MDL Number MFCD00020104
InChI Key OWGJODGIHAHWQP-KMUHKHSISA-N
Synonym N,N′C-Bis(2-hydroxybenzylidene)-1,2-propanediamine, N,N′C-Disalicylal-1,2-propanediamine
PubChem CID 50987505
IUPAC Name (6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]propylamino]methylidene]cyclohexa-2,4-dien-1-one
SMILES CC(CNC=C1C=CC=CC1=O)NC=C2C=CC=CC2=O

Specifications

Melting Point 48°C
Color Yellow
Quantity 25 g
Formula Weight 282.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(salicylidene)-1,2-propanediamine
EINECSNumber (3)-0513&(3)-1260
RTECSNumber GP3130000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.