Learn More
Medchemexpress LLC N-(n-butyl)thiophosphoric triamide | 94317-64-3 | MFCD00269941 | 99.8% | 167.21 g·mol⁻¹ | C4H14N3PS | 1 ML
SDP
Catalog No. 5000241464
Encompass_Preferred
N-(n-Butyl)thiophosphoric triamide (NBPT) is a urease inhibitor used to slow enzymatic hydrolysis of urea in soil, reducing ammonia volatilization and improving nitrogen fertilizer efficiency. It binds the nickel active site of urease, delaying urea conversion to ammonium and helping to retain nitrogen in the crop root zone.
  • Potent urease inhibitor that delays urea hydrolysis.
  • Reduces ammonia volatilization from urea-based fertilizers.
  • Improves nitrogen use efficiency and crop nitrogen uptake.
  • Available as a 10 mM solution in DMSO and as solid formulations.
  • High purity suitable for agricultural research and analytical applications.
Catalog No. 50-002-41464 Supplier Medchemexpress LLC Supplier No. HY10719910MM 1ML
N-(n-Butyl)thiophosphoric triamide (NBPT) is a urease inhibitor used to slow enzymatic hydrolysis of urea in soil, reducing ammonia volatilization and improving nitrogen fertilizer efficiency. It binds the nickel active site of urease, delaying urea conversion to ammonium and helping to retain nitrogen in the crop root zone.
  • Potent urease inhibitor that delays urea hydrolysis.
  • Reduces ammonia volatilization from urea-based fertilizers.
  • Improves nitrogen use efficiency and crop nitrogen uptake.
  • Available as a 10 mM solution in DMSO and as solid formulations.
  • High purity suitable for agricultural research and analytical applications.