Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(n-Butyl)thiophosphoric triamide, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015948 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5948 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-5948 Supplier MilliporeSigma Supelco Supplier No. 9158825MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 94317-64-3
Linear Formula C4H14N3 PS
MDL Number MFCD00269941
Melting Point 56°C to 58°C
Synonym N-Butylphosphorothioic triamide; N-Butylthiophosphoric triamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 167.21
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.