Learn More
Medchemexpress LLC N,N'-Carbonyldiimidazole | 530-62-1 | 99.9% | C7H6N4O | 1 ML
SDP
Catalog No. 5000432752
Encompass_Preferred
N,N'-Carbonyldiimidazole is a highly reactive carboxylating reagent that can activate hydroxyl groups to form reactive carbonyl groups. This compound can be used as a coupling agent and a peptide synthesis reagent.
  • Highly reactive carboxylating reagent
  • Activates hydroxyl groups to form reactive carbonyl groups
  • Functions as a coupling agent
  • Functions as a peptide synthesis reagent
  • Can mediate the conjugation of ovalbumin with magnetite nanoparticles for vaccine delivery
Catalog No. 50-004-32752 Supplier Medchemexpress LLC Supplier No. HY7988010MM/1ML
N,N'-Carbonyldiimidazole is a highly reactive carboxylating reagent that can activate hydroxyl groups to form reactive carbonyl groups. This compound can be used as a coupling agent and a peptide synthesis reagent.
  • Highly reactive carboxylating reagent
  • Activates hydroxyl groups to form reactive carbonyl groups
  • Functions as a coupling agent
  • Functions as a peptide synthesis reagent
  • Can mediate the conjugation of ovalbumin with magnetite nanoparticles for vaccine delivery