Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Di-1-naphthyl-N,N'-di-2-naphthylbenzidine 98.0+%, TCI America™
SDP

Catalog No. D4855200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4855200MG 200 mg
D48551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4855200MG Supplier TCI America Supplier No. D4855200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 374592-88-8
Molecular Formula C52H36N2
Molecular Weight (g/mol) 688.874
MDL Number MFCD16038221
InChI Key RYZPDEZIQWOVPJ-UHFFFAOYSA-N
PubChem CID 16202489
IUPAC Name N-naphthalen-1-yl-N-[4-[4-[naphthalen-1-yl(naphthalen-2-yl)amino]phenyl]phenyl]naphthalen-2-amine
SMILES C1=CC=C2C=C(C=CC2=C1)N(C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC6=CC=CC=C6C=C5)C7=CC=CC8=CC=CC=C87)C9=CC=CC1=CC=CC=C19

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 688.87
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di-1-naphthyl-N,N′-di-2-naphthylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.